accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16499-88-0

IUPAC Name: 3-butoxypropan-1-aminium
Synonyms: 3-butoxypropan-1-aminium
Molecular Weight (g/mol): 132.23
Molecular Formula: C7H18NO
InChi Key: LPUBRQWGZPPVBS-UHFFFAOYSA-O
SMILES: CCCCOCCC[NH3+]
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)