Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-n-Butoxypropylamine, 98+%, Thermo Scientific™

Catalog No. AC257551000
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC257551000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC257551000 Supplier Thermo Scientific Chemicals Supplier No. 257551000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16499-88-0
Molecular Formula C7H17NO
Molecular Weight (g/mol) 131.219
MDL Number MFCD00025622
InChI Key LPUBRQWGZPPVBS-UHFFFAOYSA-N
Synonym 3-butoxypropylamine, 3-butoxypropanamine, 1-propanamine, 3-butoxy, 3-butoxy-1-propylamine, 3-n-butoxypropylamine, butoxypropylamine, 3-n-butoxy-1-propylamine, 3-butoxy propylamine, 3-butyloxypropyl amine, 3-n-butoxyaminopropane
PubChem CID 85461
IUPAC Name 3-butoxypropan-1-amine
SMILES CCCCOCCCN

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 98+%
Refractive Index 1.4255 to 1.4275 (20°C, 589nm)
Linear Formula CH3(CH2)3O(CH2)3NH2
Quantity 100 mL
Formula Weight 131.22
Percent Purity ≥98.0%
Physical Form Liquid
Chemical Name or Material 3-n-Butoxypropylamine, 98+%
Show More Show Less
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.