Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Butoxypropylamine 98.0+%, TCI America™
SDP

Catalog No. B125025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B125025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B125025ML Supplier TCI America Supplier No. B125025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16499-88-0
Molecular Formula C7H17NO
Molecular Weight (g/mol) 131.219
MDL Number MFCD00025622
InChI Key LPUBRQWGZPPVBS-UHFFFAOYSA-N
Synonym 3-butoxypropylamine, 3-butoxypropanamine, 1-propanamine, 3-butoxy, 3-butoxy-1-propylamine, 3-n-butoxypropylamine, butoxypropylamine, 3-n-butoxy-1-propylamine, 3-butoxy propylamine, 3-butyloxypropyl amine, 3-n-butoxyaminopropane
PubChem CID 85461
IUPAC Name 3-butoxypropan-1-amine
SMILES CCCCOCCCN

Specifications

Color Yellow
Boiling Point 170°C
Quantity 25 mL
UN Number 2735
Formula Weight 131.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Butoxypropylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.