accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNOCH3HClHCl

CAS RN 165377-44-6

IUPAC Name: 1-[2-(3,4-dimethoxyphenyl)ethyl]-4-(3-phenylpropyl)piperazine dihydrochloride
Synonyms: cutamesine dihydrochloride
Molecular Weight (g/mol): 441.44
Molecular Formula: C23H34Cl2N2O2
InChi Key: XWOXAKBQEMQMFH-UHFFFAOYSA-N
SMILES: Cl.Cl.COC1=C(OC)C=C(CCN2CCN(CCCC3=CC=CC=C3)CC2)C=C1
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)