CAS RN 166518-60-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ Avasimibe
ACAT inhibitor
2
Tocris Bioscience™ Avasimibe
ACAT inhibitor