Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Avasimibe
GREENER_CHOICESDP

Catalog No. 650525 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-052-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 65-052-5 Supplier Tocris Bioscience™ Supplier No. 6505/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

ACAT inhibitor

Avasimibe is an ACAT inhibitor (IC50 = 3.3 μM). Inhibits cholesterol esterification and reduces plasma cholesterol levels in rats. Also inhibits tumor growth and prolongs survival time in a melanoma mouse model and reduces tumor growth and metastasis in a pancreatic cancer mouse model.

Specifications

Chemical Name or Material 2,6-Diisopropylphenyl (2-(2,4,6-triisopropylphenyl)acetyl)sulfamate
CAS 166518-60-1
Quantity 25 mg
Molecular Formula C29H43NO4S
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.