accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16686-11-6

IUPAC Name: 2-(3-chloropropyl)-1,3-dioxolane
Synonyms: 2-(3-chloropropyl)-1,3-dioxolane
Molecular Weight (g/mol): 150.6
Molecular Formula: C6H11ClO2
InChi Key: ZBPUNVFDQXYNDY-UHFFFAOYSA-N
SMILES: ClCCCC1OCCO1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)