CAS RN 1672691-74-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ JZP 430
Potent and irreversible α/β-hydrolase domain 6 (ABHD6) inhibitor