Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ JZP 430
GREENER_CHOICESDP

Catalog No. 584450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-445-0 50 mg
58-441-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 58-445-0 Supplier Tocris Bioscience™ Supplier No. 5844/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and irreversible α/β-hydrolase domain 6 (ABHD6) inhibitor

JZP 430 is a potent and irreversible α/β-hydrolase domain 6 (ABHD6) inhibitor (IC50 = 44 nM). Exhibits ∽230-fold selectivity over fatty acid amide hydrolase (FAAH) and lysosomal acid lipase (LAL).

Specifications

Chemical Name or Material 4-(4-Morpholinyl)-1,2,5-thiadiazol-3-yl N-cyclooctyl-N-methylcarbamate
CAS 1672691-74-5
Quantity 50 mg
Target Hydrolase Inhibitors
Molecular Formula C16H26N4O3S
Formula Weight 354.47
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.