accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONH2OO

CAS RN 167869-21-8

IUPAC Name: 2-(2-amino-3-methoxyphenyl)-4H-chromen-4-one
Synonyms: 2-(2-amino-3-methoxyphenyl)chromen-4-one
Molecular Weight (g/mol): 267.28
Molecular Formula: C16H13NO3
InChi Key: QFWCYNPOPKQOKV-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1N)C1=CC(=O)C2=CC=CC=C2O1
Product Type 
  • (4)
  • (3)
Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results