CAS RN 1684386-71-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ VER 246608
Potent and selective pyruvate dehydrogenase kinase (PDK) 1/2/3/4 inhibitor