Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ VER 246608
GREENER_CHOICESDP

Catalog No. 660450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-045-0 50 mg
66-041-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 66-045-0 Supplier Tocris Bioscience™ Supplier No. 6604/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective pyruvate dehydrogenase kinase (PDK) 1/2/3/4 inhibitor

VER 246608 is a potent and ATP-competitive inhibitor of pyruvate dehydrogenase kinase (PDK; IC50s = 35, 40, 84, and 91 nM for PDK-1, PDK-3, PDK-2, and PDK-4 respectively). It is selective for PDKs over a panel of 97 kinases and HSP90. VER 246608 increases pyruvate dehydrogenase complex activity, oxygen consumption and reduces glycolytic activity; it reduces the production of L-lactate in PC3 cells in D-glucose and L-glutamine depleted media. In PC3 cells, VER 246608 induces cell cycle arrest at the G1 phase under glucose or serum-depleted conditions characteristic of the tumor microenvironment.

Specifications

Chemical Name or Material N-[4-(2-Chloro-5-methyl-4-pyrimidinyl)phenyl]-N-[[4-[[(2,2-difluoroacetyl)amino]methyl]phenyl]methyl]-2,4-dihydroxybenzamide
CAS 1684386-71-7
Quantity 50 mg
Target Other Kinase Inhibitors
Molecular Formula C28H23ClF2N4O4
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.