CAS RN 16873-13-5
CAS RN 16873-13-5
IUPAC Name:
tetraethylazanium hydrogen sulfate
Synonyms:
tetraethylammonium hydrogen sulfate
Molecular Weight (g/mol):
227.32
Molecular Formula:
C8H21NO4S
InChi Key:
CREVBWLEPKAZBH-UHFFFAOYSA-M
SMILES:
OS([O-])(=O)=O.CC[N+](CC)(CC)CC
1
–
1
of
1
results
1
Tetraethylammonium hydrogensulfate, 99+%, HPLC grade
CAS: 16873-13-5 Molecular Formula: C8H21NO4S Molecular Weight (g/mol): 227.32 MDL Number: MFCD00036150 InChI Key: CREVBWLEPKAZBH-UHFFFAOYSA-M Synonym: tetraethylammonium hydrogensulfate,tetraethylammonium hydrogen sulphate,tetraethylammonium hydrogen sulfate,n,n,n-triethylethanaminium hydrogen sulfate 1:1:1,hydrogen sulfate; tetraethylazanium,tertraethylammonium hydrogen sulfate,tetraethyl ammonium hydrogen sulphate PubChem CID: 167582 IUPAC Name: hydrogen sulfate;tetraethylazanium SMILES: OS([O-])(=O)=O.CC[N+](CC)(CC)CC