accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16894-69-2

BrSb

CAS RN 16894-69-2

IUPAC Name: tetraphenylstibanylium bromide
Synonyms: tetraphenylstibonium bromide
Molecular Weight (g/mol): 510.09
Molecular Formula: C24H20BrSb
InChi Key: MRSXZVCXTGNNFI-UHFFFAOYSA-M
SMILES: [Br-].C1=CC=C(C=C1)[Sb+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)