accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOHOCH3H3CCH3

CAS RN 170355-37-0

IUPAC Name: 5-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydroanthracen-2-yl)thiophene-3-carboxylic acid
Synonyms: 5-(5,5,8,8-tetramethyl-6,7-dihydroanthracen-2-yl)thiophene-3-carboxylic acid
Molecular Weight (g/mol): 364.5
Molecular Formula: C23H24O2S
InChi Key: URUSABQSUCBGGJ-UHFFFAOYSA-N
SMILES: CC1(C)CCC(C)(C)C2=C1C=C1C=CC(=CC1=C2)C1=CC(=CS1)C(O)=O
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results