CAS RN 170449-18-0
IUPAC Name:
hydrogen N-(3-chlorophenyl)-6,7-dimethoxyquinazolin-4-amine chloride
Synonyms:
hydrogen N-(3-chlorophenyl)-6,7-dimethoxyquinazolin-4-amine chloride
Molecular Weight (g/mol):
352.22
Molecular Formula:
C16H15Cl2N3O2
InChi Key:
WDJDYIUSDDVWKB-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].COC1=C(OC)C=C2C(NC3=CC=CC(Cl)=C3)=NC=NC2=C1
1
–
1
of
1
results
1
R&D Systems™ AG 1478 hydrochloride
CAS: 170449-18-0 Molecular Formula: C16H15Cl2N3O2