accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNHNCH3OOOHO(E/Z)

CAS RN 171009-07-7

IUPAC Name: 6-[4-(2,3-dihydro-1,4-benzodioxin-6-yl)-5-methyl-2,3-dihydro-1H-pyrazol-3-ylidene]-4-ethyl-3-hydroxycyclohexa-2,4-dien-1-one
Synonyms: 6-[4-(2,3-dihydro-1,4-benzodioxin-6-yl)-5-methyl-1,2-dihydropyrazol-3-ylidene]-4-ethyl-3-hydroxycyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 352.39
Molecular Formula: C20H20N2O4
InChi Key: BBTMGPQSLSYPHE-UHFFFAOYSA-N
SMILES: CCC1=CC(=C2NNC(C)=C2C2=CC3=C(OCCO3)C=C2)C(=O)C=C1O
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results