accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNNNNHNBr

CAS RN 171179-06-9

IUPAC Name: N4-(3-bromophenyl)-N6-methylpyrido[3,4-d]pyrimidine-4,6-diamine
Synonyms: N4-(3-bromophenyl)-N6-methylpyrido[3,4-d]pyrimidine-4,6-diamine
Molecular Weight (g/mol): 330.19
Molecular Formula: C14H12BrN5
InChi Key: KFHMLBXBRCITHF-UHFFFAOYSA-N
SMILES: CNC1=NC=C2N=CN=C(NC3=CC(Br)=CC=C3)C2=C1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results