accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOO

CAS RN 1716-12-7

IUPAC Name: sodium 4-phenylbutanoate
Synonyms: sodium phenylbutyrate
Molecular Weight (g/mol): 186.19
Molecular Formula: C10H11NaO2
InChi Key: VPZRWNZGLKXFOE-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)CCCC1=CC=CC=C1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results