CAS RN 172889-27-9
IUPAC Name:
1-tert-butyl-3-(4-chlorophenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Synonyms:
PP2 (kinase inhibitor)
Molecular Weight (g/mol):
301.78
Molecular Formula:
C15H16ClN5
InChi Key:
PBBRWFOVCUAONR-UHFFFAOYSA-N
SMILES:
CC(C)(C)N1N=C(C2=C(N)N=CN=C12)C1=CC=C(Cl)C=C1
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ PP2
PP2, CAS 172889-27-9, is a potent, reversible, ATP-competitive, inhibitor of the Src family of protein tyrosine kinases (IC50 = 4, 5, 5, & 100 nM for p56lck, p59fynT, Hck, & Src, respectively).
2
Tocris Bioscience™ PP 2
CAS: 172889-27-9 Molecular Formula: C15H16ClN5