accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNNNO

CAS RN 17318-31-9

IUPAC Name: 9-(oxolan-2-yl)-9H-purin-6-amine
Synonyms: 9-(oxolan-2-yl)purin-6-amine
Molecular Weight (g/mol): 205.22
Molecular Formula: C9H11N5O
InChi Key: UKHMZCMKHPHFOT-UHFFFAOYNA-N
SMILES: NC1=C2N=CN(C3CCCO3)C2=NC=N1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results