accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17455-25-3

IUPAC Name: 2,5,8,11,14,21,24,27,30,33-decaoxatricyclo[32.4.0.0¹⁵,²⁰]octatriaconta-1(38),15,17,19,34,36-hexaene
Synonyms: 2,5,8,11,14,21,24,27,30,33-decaoxatricyclo[32.4.0.0¹⁵,²⁰]octatriaconta-1(38),15,17,19,34,36-hexaene
Molecular Weight (g/mol): 536.62
Molecular Formula: C28H40O10
InChi Key: MXCSCGGRLMRZMF-UHFFFAOYSA-N
SMILES: C1COCCOC2=CC=CC=C2OCCOCCOCCOCCOC2=CC=CC=C2OCCOCCO1
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results