Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzo-30-crown 10-Ether 97.0+%, TCI America™
SDP

Catalog No. D26841G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26841G Supplier TCI America Supplier No. D26841G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17455-25-3
Molecular Formula C28H40O10
Molecular Weight (g/mol) 536.62
MDL Number MFCD00054624
InChI Key MXCSCGGRLMRZMF-UHFFFAOYSA-N
Synonym dibenzo-30-crown-10, dibenzo-30-crown 10-ether, dibenzo-30-crown, dibenzo-30-crown-10-ether, 2,3,17,18-dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriaconta-2,17-diene
PubChem CID 87120
IUPAC Name 2,5,8,11,14,21,24,27,30,33-decaoxatricyclo[32.4.0.0¹⁵,²⁰]octatriaconta-1(38),15,17,19,34,36-hexaene
SMILES C1COCCOC2=CC=CC=C2OCCOCCOCCOCCOC2=CC=CC=C2OCCOCCO1

Specifications

Melting Point 107°C
Color White
Quantity 1 g
Formula Weight 536.62
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzo-30-crown 10-Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.