accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONCH3OHNCl

CAS RN 1746-81-2

IUPAC Name: 1-(4-chlorophenyl)-3-methoxy-3-methylurea
Synonyms: monolinuron
Molecular Weight (g/mol): 214.65
Molecular Formula: C9H11ClN2O2
InChi Key: LKJPSUCKSLORMF-UHFFFAOYSA-N
SMILES: CON(C)C(=O)NC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results