Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monolinuron Solution, 1000μg/mL in Acetonitrile, Ultra Scientific

Catalog No. USEPA1188
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USEPA1188 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USEPA1188 Supplier Agilent Technologies Supplier No. EPA11881
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1746-81-2 , 75-05-8
Molecular Formula C9H11ClN2O2
Molecular Weight (g/mol) 214.649
InChI Key LKJPSUCKSLORMF-UHFFFAOYSA-N
PubChem CID 15629
ChEBI CHEBI:81965
IUPAC Name 3-(4-chlorophenyl)-1-methoxy-1-methylurea
SMILES CN(C(=O)NC1=CC=C(C=C1)Cl)OC

Specifications

Physical Form Liquid
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Monolinuron, 1000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.