accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHONH3C

CAS RN 174635-69-9

IUPAC Name: 3-methyl-2-phenyl-N-(1-phenylpropyl)quinoline-4-carboxamide
Synonyms: 3-methyl-2-phenyl-N-(1-phenylpropyl)quinoline-4-carboxamide
Molecular Weight (g/mol): 380.49
Molecular Formula: C26H24N2O
InChi Key: MQNYRKWJSMQECI-UHFFFAOYNA-N
SMILES: CCC(NC(=O)C1=C2C=CC=CC2=NC(=C1C)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results