CAS RN 175178-82-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ AG 1478
A cell-permeable, reversible, ATP-competitive, highly potent and selective inhibitor of epidermal growth factor receptor kinase versus HER2-neu and platelet-derived growth factor receptor kinase.
2
MilliporeSigma™ Calbiochem™ InSolution™ AG 1478
Highly potent and specific inhibitor of the epidermal growth factor (EGF) receptor tyrosine kinase (IC50 = 3nM)