accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3OFFF

CAS RN 175205-64-8

IUPAC Name: [2-(trifluoromethoxy)phenyl]methanaminium
Synonyms: [2-(trifluoromethoxy)phenyl]methanaminium
Molecular Weight (g/mol): 192.16
Molecular Formula: C8H9F3NO
InChi Key: FYXMMFFZMQMXCQ-UHFFFAOYSA-O
SMILES: [NH3+]CC1=CC=CC=C1OC(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results