CAS RN 175883-62-2
CAS RN 175883-62-2
IUPAC Name:
(4-methoxy-3-methylphenyl)boronic acid
Synonyms:
4-methoxy-3-methylphenylboronic acid
Molecular Weight (g/mol):
165.98
Molecular Formula:
C8H11BO3
InChi Key:
PXVDQGVAZBTFIB-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1C)B(O)O
1
–
1
of
1
results
1
Thermo Scientific Chemicals 4-Methoxy-3-methylphenylboronic acid, 97%, Thermo Scientific™
CAS: 175883-62-2 Molecular Formula: C8H11BO3 Molecular Weight (g/mol): 165.98 MDL Number: MFCD02179464 InChI Key: PXVDQGVAZBTFIB-UHFFFAOYSA-N Synonym: 4-methoxy-3-methylbenzeneboronic acid,4-methoxy-3-methylphenyl boronic acid,4-methoxy-3-methylphenyl boranediol,3-methyl-4-methoxyphenylboronic acid,boronic acid, 4-methoxy-3-methylphenyl,boronic acid,b-4-methoxy-3-methylphenyl,b-4-methoxy-3-methylphenyl boronic acid,akos brn-0220,4-methoxy-3-methyl-phenyl boronic acid,timtec-bb sbb005903 PubChem CID: 2773487 IUPAC Name: (4-methoxy-3-methylphenyl)boronic acid SMILES: COC1=CC=C(C=C1C)B(O)O