accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 176199-48-7

IUPAC Name: (1S,2S,5R,6S)-2-aminobicyclo[3.1.0]hexane-2,6-dicarboxylic acid
Synonyms: eglumegad eglumetad
Molecular Weight (g/mol): 185.18
Molecular Formula: C8H11NO4
InChi Key: VTAARTQTOOYTES-RGDLXGNYSA-N
SMILES: N[C@]1(CC[C@H]2[C@@H]([C@@H]12)C(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results