Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ LY 354740
GREENER_CHOICESDP

Catalog No. 324650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-465-0 50 mg
32-461-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-465-0 Supplier R&D Systems™ Supplier No. 3246/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and highly selective group II mGlu agonist

Highly selective and potent agonist of group II mGlu receptors (EC50 values are 5.1 and 24.3 nM at mGlu2 and mGlu3 receptors respectively and > 100000 nM at mGlu4, mGlu7, mGlu1a and mGlu5a receptors). Displays antianxiety and antiaddictive activity in vivo. Orally and systemically active.

Chemical Identifiers

CAS 176199-48-7
Molecular Formula C8H11NO4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 185.18
Purity >98%
Molecular Weight (g/mol) 185.18
Chemical Name or Material LY 354740
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.