accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOHONH(E/Z)

CAS RN 177610-87-6

IUPAC Name: 6-(hydroxyimino)-N-phenyl-2-oxatricyclo[5.4.0.0³,⁵]undeca-1(11),7,9-triene-3-carboxamide
Synonyms: phccc
Molecular Weight (g/mol): 294.31
Molecular Formula: C17H14N2O3
InChi Key: FPXPIEZPAXSELW-UHFFFAOYNA-N
SMILES: ON=C1C2CC2(OC2=CC=CC=C12)C(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)