accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17805-69-5

OPOOOH2OH2OH2OCa

CAS RN 17805-69-5

IUPAC Name: calcium trihydrate naphthalen-1-yl phosphate
Synonyms: calcium trihydrate naphthalen-1-yl phosphate
Molecular Weight (g/mol): 316.26
Molecular Formula: C10H13CaO7P
InChi Key: XGDPLNLBXXLFDL-UHFFFAOYSA-L
SMILES: O.O.O.[Ca++].[O-]P([O-])(=O)OC1=CC=CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results