CAS RN 1782235-14-6
IUPAC Name:
6-[(2-{[4-(2,4-dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)pyrimidin-2-yl]amino}ethyl)amino]pyridine-3-carbonitrile trihydrochloride
Synonyms:
6-[(2-{[4-(2,4-dichlorophenyl)-5-(4-methyl-3H-imidazol-2-yl)pyrimidin-2-yl]amino}ethyl)amino]pyridine-3-carbonitrile trihydrochloride
Molecular Weight (g/mol):
574.72
Molecular Formula:
C22H21Cl5N8
InChi Key:
DSFVSCNMMZRCIA-UHFFFAOYSA-N
SMILES:
Cl.Cl.Cl.CC1=CN=C(N1)C1=CN=C(NCCNC2=NC=C(C=C2)C#N)N=C1C1=C(Cl)C=C(Cl)C=C1
1
–
1
of
1
results
1
R&D Systems™ CHIR 99021 trihydrochloride
CAS: 1782235-14-6 Molecular Formula: C22H21Cl5N8