Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CHIR 99021 trihydrochloride
GSA_VAGREENER_CHOICESDP

Catalog No. 495350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-535-0 50 mg
49-531-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 49-535-0 Supplier R&D Systems™ Supplier No. 4953/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Hydrochloride salt of CHIR 99021 (Cat. No. 4423); selective GSK-3 inhibitor

Hydrochloride salt of CHIR 99021 (Cat. No. 4423), a highly selective GSK-3 inhibitor.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 1782235-14-6
Molecular Formula C22H21Cl5N8

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 574.72
Purity >99%
Molecular Weight (g/mol) 574.72
Chemical Name or Material CHIR 99021 trihydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.