accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1782531-99-0

IUPAC Name: 5-{[3-(dimethylamino)propyl]imino}-3,10-dimethyl-1H,2H,3H,4H,5H,10H-pyrimido[4,5-b]quinoline-2,4-dione dihydrochloride
Synonyms: 5-{[3-(dimethylamino)propyl]imino}-3,10-dimethyl-1H-pyrimido[4,5-b]quinoline-2,4-dione dihydrochloride
Molecular Weight (g/mol): 414.33
Molecular Formula: C18H25Cl2N5O2
InChi Key: MADKBDHYUMEAOS-UHFFFAOYSA-N
SMILES: Cl.Cl.CN(C)CCCN=C1C2=CC=CC=C2N(C)C2=C1C(=O)N(C)C(=O)N2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results