CAS RN 180046-99-5
IUPAC Name:
N2-{1-[benzyl(methyl)carbamoyl]-2-(naphthalen-2-yl)ethyl}-N1-(2-nitrophenyl)pyrrolidine-1,2-dicarboxamide
Synonyms:
N2-{1-[benzyl(methyl)carbamoyl]-2-(naphthalen-2-yl)ethyl}-N1-(2-nitrophenyl)pyrrolidine-1,2-dicarboxamide
Molecular Weight (g/mol):
579.66
Molecular Formula:
C33H33N5O5
InChi Key:
PCVSIMQAFWRUEC-UHFFFAOYNA-N
SMILES:
CN(CC1=CC=CC=C1)C(=O)C(CC1=CC=C2C=CC=CC2=C1)NC(=O)C1CCCN1C(=O)NC1=CC=CC=C1[N+]([O-])=O