CAS RN 180605-36-1
CAS RN 180605-36-1
IUPAC Name:
4-(2-aminophenyl)-1-methylpiperazin-1-ium
Synonyms:
4-(2-aminophenyl)-1-methylpiperazin-1-ium
Molecular Weight (g/mol):
192.29
Molecular Formula:
C11H18N3
InChi Key:
INWHDRNGZMHXEZ-UHFFFAOYSA-O
SMILES:
C[NH+]1CCN(CC1)C1=CC=CC=C1N
1
–
1
of
1
results
1
2-(4-Methyl-1-piperazinyl)aniline 98.0+%, TCI America™

CAS: 180605-36-1 Molecular Formula: C11H17N3 Molecular Weight (g/mol): 191.278 MDL Number: MFCD04035359 InChI Key: INWHDRNGZMHXEZ-UHFFFAOYSA-N Synonym: 2-4-methylpiperazin-1-yl aniline,2-4-methyl-1-piperazinyl aniline,2-4-methylpiperazino aniline,2-4-methyl-piperazin-1-yl-phenylamine,benzenamine, 2-4-methyl-1-piperazinyl,1-2-aminophenyl-4-methylpiperazine,2-4-methylpiperazinyl phenylamine,asischem z51464,pubchem13000,akos bb-7895 PubChem CID: 286547 IUPAC Name: 2-(4-methylpiperazin-1-yl)aniline SMILES: CN1CCN(CC1)C2=CC=CC=C2N