accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOHOSNHNH2ONOO

CAS RN 182004-65-5

IUPAC Name: [(2-{4-[(4-nitrophenyl)methoxy]phenyl}ethyl)sulfanyl]methanimidamide; methanesulfonic acid
Synonyms: (2-{4-[(4-nitrophenyl)methoxy]phenyl}ethyl)sulfanylmethanimidamide; methanesulfonic acid
Molecular Weight (g/mol): 427.49
Molecular Formula: C17H21N3O6S2
InChi Key: WGIKEBHIKKWJLG-UHFFFAOYSA-N
SMILES: CS(O)(=O)=O.NC(=N)SCCC1=CC=C(OCC2=CC=C(C=C2)[N+]([O-])=O)C=C1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)