CAS RN 18213-75-7
CAS RN 18213-75-7
IUPAC Name:
5-amino-1-methyl-1H-pyrazole-4-carboxamide
Synonyms:
5-amino-1-methylpyrazole-4-carboxamide
Molecular Weight (g/mol):
140.15
Molecular Formula:
C5H8N4O
InChi Key:
JGSQVTVXGXOSCH-UHFFFAOYSA-N
SMILES:
CN1N=CC(C(N)=O)=C1N
1
–
1
of
1
results
1
5-Amino-1-methyl-1H-pyrazole-4-carboxamide, Thermo Scientific™
CAS: 18213-75-7 Molecular Formula: C5H8N4O Molecular Weight (g/mol): 140.146 MDL Number: MFCD00111808 InChI Key: JGSQVTVXGXOSCH-UHFFFAOYSA-N PubChem CID: 265696 IUPAC Name: 5-amino-1-methylpyrazole-4-carboxamide SMILES: CN1C(=C(C=N1)C(=O)N)N