accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 185077-02-5

CH3FMgBr

CAS RN 185077-02-5

IUPAC Name: bromo(3-fluoro-4-methylphenyl)magnesium
Synonyms: bromo(3-fluoro-4-methylphenyl)magnesium
Molecular Weight (g/mol): 213.33
Molecular Formula: C7H6BrFMg
InChi Key: FGTAIRVIEJUZJO-UHFFFAOYSA-M
SMILES: CC1=CC=C([Mg]Br)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results