CAS RN 185246-32-6
IUPAC Name:
2-bromo-3-{2,7-dihydroxy-2-sulfanylidene-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-6-yl}-6-phenyl-3H,5H,9H-imidazo[1,2-a]purin-9-one sodium
Synonyms:
2-bromo-3-{2,7-dihydroxy-2-sulfanylidene-tetrahydro-4H-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-6-yl}-6-phenyl-5H-imidazo[1,2-a]purin-9-one sodium
Molecular Weight (g/mol):
563.27
Molecular Formula:
C18H15BrN5NaO6PS
InChi Key:
HHYKGBCHGONJCV-UHFFFAOYNA-N
SMILES:
[Na].OC1C(OC2COP(O)(=S)OC12)N1C(Br)=NC2=C1N=C1NC(=CN1C2=O)C1=CC=CC=C1
1
–
1
of
1
results
1
R&D Systems™ Rp-8-Br-PET-cGMPS
CAS: 185246-32-6 Molecular Formula: C18H15BrN5NaO6PS