Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Rp-8-Br-PET-cGMPS
GREENER_CHOICESDP

Catalog No. 3028560U Shop All Novus Products
Change view
Click to view available options
Quantity:
560 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-285-60U 560 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 30-285-60U Supplier R&D Systems™ Supplier No. 3028/560U

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Protein kinase G inhibitor

Competitive, reversible cGMP-dependent protein kinase (PKG) inhibitor; cGMP analog.

This product is sold in units of 560μg, equivalent to 1μmol.

Chemical Identifiers

CAS 185246-32-6
Molecular Formula C18H15BrN5NaO6PS

Specifications

Quantity 560 μg
Product Line Tocris
Formula Weight 562.27
Purity >99%
Molecular Weight (g/mol) 563.275
Chemical Name or Material Rp-8-Br-PET-cGMPS
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.