CAS RN 185991-07-5
IUPAC Name:
[(pyridin-2-yl)methyl]({4-[(1,4,8,11-tetraazacyclotetradecan-1-yl)methyl]phenyl}methyl)amine
Synonyms:
(pyridin-2-ylmethyl)({[4-(1,4,8,11-tetraazacyclotetradecan-1-ylmethyl)phenyl]methyl})amine
Molecular Weight (g/mol):
410.61
Molecular Formula:
C24H38N6
InChi Key:
CWJJHESJXJQCJA-UHFFFAOYSA-N
SMILES:
C(NCC1=CC=CC=N1)C1=CC=C(CN2CCCNCCNCCCNCC2)C=C1
1
–
1
of
1
results
1
R&D Systems™ AMD 3465 hexahydrobromide
CAS: 185991-07-5 Molecular Formula: C24H44Br6N6