Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AMD 3465 hexahydrobromide
GREENER_CHOICESDP

Catalog No. 417950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-795-0 50 mg
41-791-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-795-0 Supplier R&D Systems™ Supplier No. 4179/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective CXCR4 antagonist

Potent, selective CXCR4 antagonist; exhibits 8-fold higher affinity than AMD 3100 (Cat.No. 3299). Inhibits SDF-1α-ligand binding (Ki = 41.7 nM). Potently inhibits HIV cell entry in vitro; causes leukocytosis and mobilizes haematopoietic stem cells in vivo.

Chemical Identifiers

CAS 185991-07-5
Molecular Formula C24H44Br6N6

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 896.07
Purity >97%
Molecular Weight (g/mol) 896.082
Chemical Name or Material AMD 3465 hexahydrobromide
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.