accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOONNOCH3HCl

CAS RN 186002-54-0

IUPAC Name: 1-(ethanesulfonyl)-10-methoxy-2,3,4,4a,5,7,8,12b,13,13a-decahydro-1H-1,6-diazatetraphene hydrochloride
Synonyms: 1-(ethanesulfonyl)-10-methoxy-2,3,4,4a,5,7,8,12b,13,13a-decahydro-1,6-diazatetraphene hydrochloride
Molecular Weight (g/mol): 400.96
Molecular Formula: C19H29ClN2O3S
InChi Key: DZTZUOBWDBPPJQ-UHFFFAOYNA-N
SMILES: Cl.CCS(=O)(=O)N1CCCC2CN3CCC4=C(C=CC(OC)=C4)C3CC12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)