accessibility menu, dialog, popup

UserName

Viewing profile as user:
NONNOOHNOOHOHOHOH

CAS RN 186689-07-6

IUPAC Name: 3,4,5,6-tetrahydroxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]hexanal
Synonyms: 3,4,5,6-tetrahydroxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]hexanal
Molecular Weight (g/mol): 342.26
Molecular Formula: C12H14N4O8
InChi Key: QUTFFEUUGHUPQC-UHFFFAOYNA-N
SMILES: OCC(O)C(O)C(O)C(NC1=CC=C(C2=NON=C12)[N+]([O-])=O)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results