Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ 2-NBDG
GREENER_CHOICESDP

Catalog No. 60655 Shop All Novus Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-655 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 60-655 Supplier R&D Systems™ Supplier No. 6065/5

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Fluorescent glucose uptake indicator

Fluorescent glucose analog for visualizing glucose uptake into living cells.

Chemical Identifiers

CAS 186689-07-6
Molecular Formula C12H14N4O8

Specifications

Quantity 5 mg
Solubility Soluble to 100mM in DMSO
Product Line Tocris
Formula Weight 342.26
Physical Form Solid
Purity >97%
Molecular Weight (g/mol) 342.264
Chemical Name or Material 2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.