CAS RN 1867107-62-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ RA 2
Potent negative modulator of KCa2.3 and KCa3.1 channels