accessibility menu, dialog, popup

UserName

Viewing profile as user:
NONHHOClCl

CAS RN 18711-16-5

IUPAC Name: 6,7-dichloro-3-(hydroxyamino)-2H-indol-2-one
Synonyms: 6,7-dichloro-3-(hydroxyamino)indol-2-one
Molecular Weight (g/mol): 231.03
Molecular Formula: C8H4Cl2N2O2
InChi Key: CVOUSAVHMDXCKG-UHFFFAOYSA-N
SMILES: ONC1=C2C=CC(Cl)=C(Cl)C2=NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results